Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mat2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145569.4 | NP_663544.1 | 9 | 1388 | ||
GenBank mRNA | AB070266.1 | - | 9 | 1352 | ||
AK034757.1 | - | 9 | 1389 | |||
AK052764.1 | - | 9 | 1387 | |||
AK079420.1 | - | 9 | 1394 | |||
AK144189.1 | - | 9 | 1391 | |||
AK145407.1 | - | 9 | 1387 | |||
AK150040.1 | - | 9 | 1387 | |||
AK151281.1 | - | 8 | 1973 | |||
AK160869.1 | - | 8 | 1972 | |||
AK161852.1 | - | 8 | 1971 | |||
AK164137.1 | - | 9 | 1387 | |||
AK166671.1 | - | 9 | 1387 | |||
AK168155.1 | - | 9 | 1387 | |||
AK169705.1 | - | 9 | 1387 | |||
AK210966.1 | - | 3 | 371 | |||
AK213373.1 | - | 1 | 77 | |||
AK215700.1 | - | 1 | 89 | |||
BC003451.1 | - | 9 | 1328 | 2655444 | ||
BC058360.1 | - | 8 | 1949 | 6821781 |