Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145610.2 | NP_663585.2 | 11 | 1533 | ||
GenBank mRNA | AK039027.1 | - | 11 | 1438 | ||
AK085695.1 | - | 1 | 3777 | |||
AK171672.1 | - | 7 | 1199 | |||
AK172615.1 | - | 7 | 1199 | |||
BB749945.1 | - | 2 | 298 | |||
BC014688.1 | - | 11 | 1451 | 4489442 | ||
BC059277.1 | - | 9 | 1697 |