Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145611.4 | NP_663586.3 | 12 | 3523 | ||
GenBank mRNA | AK031324.1 | - | 12 | 3516 | ||
AK045492.1 | - | 12 | 3493 | |||
AK202086.1 | - | 1 | 254 | |||
BC010245.1 | - | 11 | 3386 | 3710765 |