Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145636.1 | NP_663611.1 | 5 | 471 | ||
GenBank mRNA | AK152114.1 | - | 5 | 493 | ||
AK153466.1 | - | 5 | 493 | |||
AY099297.1 | - | 5 | 471 | |||
BC119402.1 | - | 5 | 502 | 8734151 |