Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgcs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291439.1 | NP_001278368.1 | 2 | 520 | ||
NM_145942.5 | NP_666054.2 | 3 | 579 | |||
GenBank mRNA | AK031297.1 | - | 3 | 468 | ||
AK044835.1 | - | 3 | 469 | |||
AK045094.1 | - | 2 | 408 | |||
AK078743.1 | - | 2 | 410 | |||
AK135551.1 | - | 2 | 408 | |||
AK136171.1 | - | 3 | 468 | |||
AK144384.1 | - | 2 | 408 | |||
AK145424.1 | - | 2 | 408 | |||
AK146042.1 | - | 2 | 410 | |||
AK146590.1 | - | 2 | 410 | |||
BC023851.1 | - | 3 | 495 | 5347038 | ||
BC029693.1 | - | 2 | 425 | 5375374 | ||
BC031363.1 | - | 2 | 427 | 5357048 | ||
BC034317.1 | - | 1 | 371 | 5366786 |