Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013254.3 | NP_037386.1 | 17 | 1997 | ||
XM_005268809.1 | XP_005268866.1 | 17 | 2075 | |||
XM_005268810.1 | XP_005268867.1 | 17 | 2057 | |||
GenBank mRNA | AF174536.1 | - | 16 | 1838 | ||
AF191838.1 | - | 17 | 1900 | |||
AK002192.1 | - | 9 | 823 | |||
AK291039.1 | - | 17 | 1942 | |||
AK303683.1 | - | 12 | 1214 | |||
BC034950.2 | - | 17 | 1937 | 4824988 |