Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc45a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177628.2 | NP_001171099.1 | 2 | 516 | ||
NM_145977.2 | NP_666089.1 | 2 | 597 | |||
GenBank mRNA | AK035428.1 | - | 2 | 466 | ||
AK162123.1 | - | 2 | 444 | |||
BC031381.1 | - | 2 | 597 | 5050610 | ||
BY713333.1 | - | 2 | 493 |