Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145824.4 | NP_665823.2 | 7 | 1041 | ||
GenBank mRNA | AK173177.1 | - | 7 | 854 | ||
AY337314.1 | - | 7 | 1042 | |||
BC024698.1 | - | 7 | 886 | 3667000 | ||
EU281316.1 | - | 7 | 902 |