Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St6gal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252505.1 | NP_001239434.1 | 8 | 3949 | ||
NM_001252506.1 | NP_001239435.1 | 7 | 3816 | |||
NM_145933.4 | NP_666045.1 | 8 | 4280 | |||
GenBank mRNA | AK034768.1 | - | 2 | 2626 | ||
AK084124.1 | - | 8 | 4115 | |||
BC027833.1 | - | 5 | 3477 | 5321763 | ||
BC092222.1 | - | 8 | 4258 | 6844069 | ||
BC096026.1 | - | 8 | 3925 | 6833540 | ||
CJ238175.1 | - | 1 | 467 |