Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glyat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145935.3 | NP_666047.1 | 5, 6 | 804 | ||
GenBank mRNA | AK039262.1 | - | 5, 6 | 805 | ||
AK143803.1 | - | 594 | ||||
AK143901.1 | - | 594 | ||||
BC010799.1 | - | 5, 6 | 579 | 4234793 | ||
BC015294.1 | - | 616 | 4220471 | |||
BC024434.1 | - | 4, 5 | 494 | 4972912 |