Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk5rap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145990.3 | NP_666102.2 | 35 | 5451 | ||
GenBank mRNA | AK129411.1 | - | 35 | 5451 | ||
BC027761.1 | - | 1248 | ||||
BC059253.1 | - | 23 | 4042 | |||
BC079888.1 | - | 32 | 5095 | |||
BC086696.1 | - | 35 | 5353 | 30691910 | ||
BC110364.1 | - | 7 | 983 |