Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zadh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146090.5 | NP_666202.2 | 2 | 853 | ||
GenBank mRNA | AK047559.1 | - | 2 | 858 | ||
AK081988.1 | - | 2 | 854 | |||
AK083082.1 | - | 2 | 897 | |||
BC025897.1 | - | 1 | 94 | |||
BC116427.1 | - | 2 | 854 | 40104327 | ||
BC116428.1 | - | 2 | 854 | 40104329 |