Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177798.1 | NP_001171269.1 | 4, 5 | 561 | ||
NM_146092.2 | NP_666204.2 | 3, 4 | 323 | |||
GenBank mRNA | AK077149.1 | - | 4, 5 | 562 | ||
AK144881.1 | - | 3, 4 | 290 | |||
AK178817.1 | - | 1, 2 | 95 | |||
BC020299.1 | - | 3, 4 | 296 | |||
BC028647.1 | - | 3, 4 | 270 | 1244243 |