Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B4galt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146045.1 | NP_666157.1 | 2, 3 | 439 | ||
GenBank mRNA | AK050684.1 | - | 2, 3 | 386 | ||
AK141739.1 | - | 2, 3 | 403 | |||
AK146855.1 | - | 2, 3 | 486 | |||
AK171600.1 | - | 2, 3 | 455 | |||
BC027195.1 | - | 2, 3 | 439 | 4224150 | ||
BC055703.1 | - | 2, 3 | 654 | 6403624 |