Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fermt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146054.2 | NP_666166.2 | 12 | 1579 | ||
GenBank mRNA | AK145030.1 | - | 12 | 1521 | ||
AK165608.1 | - | 12 | 1549 | |||
AK166552.1 | - | 12 | 1518 | |||
BC033436.1 | - | 12 | 1500 | 4950810 | ||
BC034168.1 | - | 3 | 233 | |||
CD550909.1 | - | 5 | 443 |