Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpped1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146067.3 | NP_666179.2 | 3, 4 | 808 | ||
GenBank mRNA | AK029379.1 | - | 3, 4 | 733 | ||
AK032288.1 | - | 3, 4 | 771 | |||
AK049721.1 | - | 3, 4 | 809 | |||
AK080876.1 | - | 3, 4 | 766 | |||
AK133977.1 | - | 3, 4 | 802 | |||
BC025476.1 | - | 3, 4 | 729 | |||
BC034347.1 | - | 3, 4 | 450 | 2654874 |