Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lemd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146075.2 | NP_666187.2 | 3, 4 | 905 | ||
GenBank mRNA | AK082131.1 | - | 3, 4 | 712 | ||
AK143224.1 | - | 3, 4 | 890 | |||
AY640307.1 | - | 3, 4 | 885 | |||
BC026588.1 | - | 3, 4 | 215 | 4972900 | ||
BC050010.1 | - | 1, 2 | 1117 |