Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177798.1 | NP_001171269.1 | 6 | 707 | ||
NM_146092.2 | NP_666204.2 | 5 | 469 | |||
GenBank mRNA | AK077149.1 | - | 6 | 708 | ||
AK144881.1 | - | 5 | 436 | |||
AK178817.1 | - | 3 | 241 | |||
BC020299.1 | - | 5 | 442 | |||
BC028647.1 | - | 5 | 416 | 1244243 |