Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctage5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146034.3 | NP_666146.3 | 5 | 409 | ||
GenBank mRNA | AK053010.1 | - | 5 | 343 | ||
AK080312.1 | - | 5 | 385 | |||
AK087188.1 | - | 4 | 324 | |||
AK208297.1 | - | 5 | 282 | |||
BC026864.1 | - | 5 | 361 | 2647214 |