Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000668.5 | NP_000659.2 | 9 | 1773 | ||
NM_001286650.1 | NP_001273579.1 | 10 | 1895 | |||
GenBank mRNA | BC033009.2 | - | 9 | 1732 | 4828235 | |
BX647469.1 | - | 9 | 3757 | |||
D00137.1 | - | 9 | 1682 | |||
FJ224285.1 | - | 9 | 1732 | |||
M21692.1 | - | 9 | 1694 | |||
X03350.1 | - | 9 | 1734 |