Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afap1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177796.1 | NP_001171267.1 | 20 | 3362 | ||
NM_001177797.1 | NP_001171268.1 | 19 | 3308 | |||
NM_146102.2 | NP_666214.1 | 21 | 3414 | |||
GenBank mRNA | AK048649.1 | - | 21 | 3068 | ||
AK087449.1 | - | 20 | 2999 | |||
AK220430.1 | - | 19 | 2931 | |||
BC031515.1 | - | 20 | 2964 | 3711746 |