Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146189.3 | NP_666301.2 | 3 | 230 | ||
GenBank mRNA | AK029282.1 | - | 3 | 216 | ||
AK132563.1 | - | 3 | 218 | |||
AK161146.1 | - | 3 | 218 | |||
BC029762.1 | - | 3 | 200 | 3983351 | ||
BC039184.1 | - | 3 | 195 | 3980667 | ||
CK787764.1 | - | 3 | 244 |