Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golph3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177670.1 | NP_001171141.1 | 2 | 307 | ||
NM_146133.3 | NP_666245.2 | 2 | 307 | |||
GenBank mRNA | AK008434.1 | - | 2 | 274 | ||
AK167675.1 | - | 2 | 257 | |||
AK169724.1 | - | 2 | 261 | |||
BC027194.1 | - | 2 | 217 | 4216692 | ||
BC047147.1 | - | 2 | 282 | 6465983 | ||
BY107376.1 | - | 2 | 308 |