Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146602.2 | NP_666813.2 | 1 | 896 | ||
GenBank mRNA | BC113955.1 | - | 1 | 895 | ||
BC148217.1 | - | 1 | 891 |