Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146600.1 | NP_666811.1 | 1 | 477 | ||
GenBank mRNA | BC104090.1 | - | 1 | 477 | ||
BC104091.2 | - | 1 | 477 | 40042950 | ||
BC106811.1 | - | 1 | 477 | |||
BC106812.2 | - | 1 | 477 | 40043224 |