Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001074.2 | NP_001065.2 | 2, 3 | 924 | ||
XM_005265702.3 | XP_005265759.1 | 3, 4 | 628 | |||
XM_011532229.2 | XP_011530531.1 | 2, 3 | 925 | |||
XM_011532230.1 | XP_011530532.1 | 2, 3 | 925 | |||
GenBank mRNA | AK223142.1 | - | 2, 3 | 898 | ||
AK313190.1 | - | 2, 3 | 925 | |||
BC030974.1 | - | 2, 3 | 924 | 4593426 | ||
J05428.1 | - | 2, 3 | 892 |