Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166556.1 | NP_001160028.1 | 2 | 301 | ||
NM_001166557.1 | NP_001160029.1 | 2 | 301 | |||
NM_147218.2 | NP_671751.2 | 2 | 301 | |||
GenBank mRNA | AF498361.1 | - | 2 | 253 | ||
AK018242.1 | - | 2 | 286 | |||
AK040652.1 | - | 2 | 301 | |||
AK042934.1 | - | 2 | 273 | |||
AK053384.1 | - | 2 | 228 | |||
BC132417.1 | - | 2 | 138 | 40130694 | ||
BC132419.1 | - | 2 | 138 | 40130696 |