Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130881.2 | NP_570951.2 | 8 | 1837 | ||
NM_148917.2 | NP_683717.1 | 8 | 1463 | |||
GenBank mRNA | AK135962.1 | - | 1302 | |||
BC010345.1 | - | 8 | 1463 | 3707766 | ||
BC056432.1 | - | 8 | 1837 | 5696369 | ||
BC058263.1 | - | 8 | 2049 |