Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148930.3 | NP_683732.1 | 3 | 317 | ||
GenBank mRNA | AJ309168.1 | - | 2 | 152 | ||
AK133999.1 | - | 3 | 275 | |||
AK134183.1 | - | 3 | 312 | |||
AK146880.1 | - | 3 | 312 | |||
AK161794.1 | - | 3 | 304 | |||
BC019766.1 | - | 3 | 266 | |||
BC023854.1 | - | 3 | 302 | 5346780 | ||
BC031899.1 | - | 3 | 302 | 5362121 | ||
BC042419.1 | - | 3 | 295 | |||
BC043058.1 | - | 3 | 254 | 6419695 | ||
BC054729.1 | - | 3 | 305 | 6419758 | ||
BC054820.1 | - | 3 | 301 | |||
BC060985.1 | - | 3 | 295 |