Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152813.3 | NP_690026.2 | 4 | 739 | ||
GenBank mRNA | AK046669.1 | - | 4 | 705 | ||
AK144950.1 | - | 1812 | ||||
AK164809.1 | - | 4 | 705 | |||
AK166977.1 | - | 4 | 732 | |||
AK173311.1 | - | 4 | 711 | |||
BC031392.1 | - | 4 | 683 | 4952407 |