Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147220.2 | NP_671753.2 | 3, 4 | 565 | ||
GenBank mRNA | AF491299.1 | - | 3, 4 | 565 |