Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK047751.1 | - | 3 | 296 | ||
AK135543.1 | - | 3 | 304 | |||
AK156828.1 | - | 3 | 301 | |||
AK180912.1 | - | 2 | 193 | |||
BC024418.1 | - | 3 | 339 | 4982610 | ||
BC026567.1 | - | 3 | 327 | 4982239 | ||
BC085273.1 | - | 3 | 279 | 30144329 | ||
CJ060196.1 | - | 3 | 341 |