Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148926.2 | NP_683728.1 | 5 | 2475 | ||
GenBank mRNA | AK082915.1 | - | 4 | 2471 | ||
AK148316.1 | - | 2505 | ||||
BC024483.1 | - | 4 | 2137 | |||
BC027161.1 | - | 4 | 2274 | |||
BC034396.1 | - | 5 | 2396 | 3707476 | ||
BC083124.1 | - | 5 | 2475 | 6334368 |