Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dner | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152915.1 | NP_690879.1 | 3631 | |||
GenBank mRNA | BC034634.1 | - | 3631 | 5365190 |