Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000982.3 | NP_000973.2 | 1 | 180 | ||
GenBank mRNA | AF086172.1 | - | 1 | 79 | ||
AK130968.1 | - | 1 | 179 | |||
AK311795.1 | - | 1 | 177 | |||
AY320407.1 | - | 1 | 180 | |||
BC001603.1 | - | 1 | 162 | 3543702 | ||
BC007505.2 | - | 1 | 143 | 2964441 | ||
BC019002.2 | - | 1 | 135 | |||
BC062981.1 | - | 1 | 160 | 6084268 | ||
BC070184.1 | - | 1 | 164 | 4281405 | ||
BC070185.1 | - | 1 | 177 | |||
BC070323.1 | - | 1 | 177 | 4248655 | ||
BC070330.1 | - | 1 | 177 | 3931616 | ||
BC071902.1 | - | 1 | 160 | 4901492 | ||
BC104667.1 | - | 1 | 181 | 6724053 | ||
BX647669.1 | - | 1 | 179 | |||
CR457032.1 | - | 1 | 137 | |||
U14967.1 | - | 1 | 166 | |||
U25789.1 | - | 1 | 170 | |||
X89401.1 | - | 1 | 150 |
Gene Symbol: RPL21P19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB209100.1 | - | 1 | 239 |