Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153133.2 | NP_694773.1 | 4 | 1549 | ||
GenBank mRNA | AB081322.1 | - | 4 | 1487 | ||
AF372838.1 | - | 4 | 1551 | |||
AK132181.1 | - | 1620 | ||||
BC141318.1 | - | 4 | 1529 | 9055930 |