Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153195.2 | NP_694875.2 | 2 | 399 | ||
GenBank mRNA | AK147797.1 | - | 2 | 351 | ||
AK147907.1 | - | 2 | 318 | |||
AK150190.1 | - | 2 | 174 | |||
AK150626.1 | - | 2 | 357 | |||
AK151025.1 | - | 2 | 357 | |||
AK152513.1 | - | 2 | 357 | |||
AK182909.1 | - | 11 | ||||
BC032153.1 | - | 2 | 341 | 5054087 | ||
CD563204.1 | - | 2 | 399 |