Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146298.2 | NP_001139770.1 | 12, 13 | 1948 | ||
NM_001282093.1 | NP_001269022.1 | 13, 14 | 2098 | |||
NM_153085.4 | NP_694725.3 | 13, 14 | 2257 | |||
GenBank mRNA | AF320996.1 | - | 13, 14 | 2037 | ||
AK044675.1 | - | 6, 7 | 930 | |||
AK156780.1 | - | 13, 14 | 2082 | |||
AK172416.1 | - | 13, 14 | 2082 | |||
AK173274.1 | - | 12, 13 | 1650 | |||
BC020146.1 | - | 12, 13 | 1748 | |||
BC057178.1 | - | 13, 14 | 1889 | |||
BC080851.1 | - | 13, 14 | 2215 | 6848738 | ||
BM948716.1 | - | 3, 4 | 411 | |||
CB524834.1 | - | 3, 4 | 417 |