Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153097.2 | NP_694737.1 | 2 | 1414 | ||
GenBank mRNA | AF290197.1 | - | 2 | 1316 | ||
AK135841.1 | - | 2 | 1311 | |||
BC139468.1 | - | 2 | 1298 | 8862490 | ||
BC139469.1 | - | 2 | 1298 | 8862491 |