Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153125.2 | NP_694765.2 | 16, 17 | 5673 | ||
GenBank mRNA | AK122242.2 | - | 9, 10 | 1110 | ||
BC034649.1 | - | 6, 7 | 731 | |||
BC042603.1 | - | 10, 11 | 1189 | |||
BC059898.1 | - | 7, 8 | 1276 |