Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001020.5 | NP_001011.1 | 5 | 439 | ||
NM_001321111.1 | NP_001308040.1 | 5 | 388 | |||
XM_005259137.2 | XP_005259194.1 | 4 | 534 | |||
GenBank mRNA | AK055869.1 | - | 4 | 2212 | ||
AK315498.1 | - | 5 | 367 | |||
BC004324.2 | - | 5 | 365 | 3628799 | ||
BC007977.2 | - | 5 | 350 | 4310307 | ||
BC024970.1 | - | 1 | 335 | |||
BC071674.1 | - | 4 | 463 | 5394209 | ||
BC095429.1 | - | 5 | 309 | |||
BQ421722.1 | - | 5 | 397 | |||
M60854.1 | - | 5 | 351 |
Gene Symbol: ZNF90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK298173.1 | - | 3 | 584 |