Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153419.2 | NP_700468.2 | 3 | 412 | ||
GenBank mRNA | AB001539.1 | - | 4 | 484 | ||
AK077611.1 | - | 3 | 418 | |||
BC008121.1 | - | 3 | 389 | |||
BC028896.1 | - | 2 | 281 | 3674738 | ||
BC083143.1 | - | 3 | 400 | |||
BQ886252.1 | - | 3 | 415 |