Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhobtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153514.5 | NP_705734.4 | 10 | 3088 | ||
GenBank mRNA | AK038196.1 | - | 10 | 3180 | ||
AK081100.1 | - | 10 | 3017 |