Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153518.1 | NP_705738.1 | 1 | 276 | ||
GenBank mRNA | AF454432.1 | - | 1 | 276 | ||
AK077174.1 | - | 1 | 310 | |||
BC048073.1 | - | 1 | 296 | 6312435 | ||
BC056947.1 | - | 1 | 272 | |||
BC060155.1 | - | 1 | 272 |