Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aaas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153416.2 | NP_700465.2 | 5 | 553 | ||
GenBank mRNA | AK034591.1 | - | 5 | 448 | ||
AK079849.1 | - | 5 | 490 | |||
AK083401.1 | - | 5 | 466 | |||
AK083537.1 | - | 5 | 474 | |||
AK087134.1 | - | 5 | 529 | |||
AK088247.1 | - | 5 | 470 | |||
AK134406.1 | - | 5 | 444 | |||
AK165243.1 | - | 5 | 468 | |||
AK166118.1 | - | 5 | 468 | |||
AK169523.1 | - | 5 | 505 | |||
AK189195.1 | - | 4 | 244 | |||
BC018191.1 | - | 5 | 454 | 3994248 | ||
BC025501.1 | - | 5 | 500 | 5321013 | ||
BC026604.1 | - | 4 | 722 |