Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293693.1 | NP_001280622.1 | 20 | 2228 | ||
NM_153397.2 | NP_700446.2 | 22 | 2428 | |||
GenBank mRNA | AF513715.1 | - | 22 | 2411 | ||
AK005759.1 | - | 20 | 2219 | |||
AV256066.1 | - | 1 | 203 | |||
BC060983.1 | - | 23 | 2425 | 6772485 |