Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153419.2 | NP_700468.2 | 7 | 1520 | ||
GenBank mRNA | AB001539.1 | - | 8 | 1591 | ||
AK077611.1 | - | 7 | 1525 | |||
AK210248.1 | - | 1 | 52 | |||
AK210821.1 | - | 11 | ||||
BC008121.1 | - | 7 | 1497 | |||
BC028896.1 | - | 6 | 1389 | 3674738 | ||
BC083143.1 | - | 7 | 1508 | |||
DT904438.1 | - | 1 | 335 |