Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2l13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153516.2 | NP_705736.1 | 7 | 2458 | ||
GenBank mRNA | AK031018.1 | - | 7 | 2407 | ||
AK047223.1 | - | 7 | 2460 | |||
AK088417.1 | - | 7 | 2473 | |||
AK132774.1 | - | 7 | 2467 | |||
AK143457.1 | - | 7 | 2474 | |||
AK148007.1 | - | 7 | 2477 | |||
AK165068.1 | - | 7 | 2452 | |||
AK168833.1 | - | 7 | 2448 | |||
BC027307.1 | - | 7 | 2348 | 4014844 | ||
BC029016.1 | - | 7 | 2458 | 5358785 |