Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slain2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113423.2 | NP_001106894.1 | 5 | 1454 | ||
NM_153567.3 | NP_705795.2 | 5 | 1454 | |||
GenBank mRNA | AK039970.1 | - | 5 | 1399 | ||
AK129365.1 | - | 5 | 1443 | |||
AK146432.1 | - | 5 | 1453 | |||
AK216525.1 | - | 1 | 39 | |||
BC010836.1 | - | 5 | 1442 | |||
BC038019.1 | - | 3 | 659 | 3597423 | ||
BC076573.1 | - | 5 | 1387 | |||
CK619668.1 | - | 1 | 161 |