Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153583.10 | NP_705811.8 | 1 | 389 | ||
GenBank mRNA | AK036282.1 | - | 1 | 376 | ||
AK036571.1 | - | 1 | 378 | |||
AK044798.1 | - | 1 | 367 | |||
AK080353.1 | - | 1 | 369 | |||
AK081002.1 | - | 1 | 376 | |||
AK081605.1 | - | 1 | 367 | |||
AK087258.1 | - | 1 | 379 | |||
AK154130.1 | - | 1 | 382 | |||
BB857077.1 | - | 1 | 391 | |||
BC030861.1 | - | 1 | 366 | 4208700 | ||
BC069851.1 | - | 1 | 341 | 6490007 |